APO455833832 Apollo Scientific Ltd

6-Chloro-2-fluoro-3-methylbenzoyl chloride

  • MDL number MFCD01631371
  • Cas Number : 261762-82-7
  • Molecular Weight : 207.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Chloro-2-fluoro-m-toluoyl chloride, 4-Chloro-3-(chlorocarbonyl)-2-fluorotoluene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110/1mm
Product Link https://store.apolloscientific.co.uk/product/6-chloro-2-fluoro-3-methylbenzoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chloro-2-fluoro-3-methylbenzoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart