APO455833834 Apollo Scientific Ltd

2-Chloro-6-fluoro-3-methylbenzyl alcohol

  • MDL number MFCD01631362
  • Cas Number : 261762-83-8
  • Molecular Weight : 174.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Synonym 2-Chloro-4-fluoro-3-(hydroxymethyl)toluene
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P264;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart