APO455833840 Apollo Scientific Ltd

2-Chloro-6-fluoro-3-methylphenylacetic acid

  • MDL number MFCD01631365
  • Cas Number : 261762-92-9
  • Molecular Weight : 202.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-fluoro-m-tolylacetic acid, 3-(Carboxymethyl)-2-chloro-4-fluorotoluene
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylphenylacetic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylphenylacetic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart