APO455833842 Apollo Scientific Ltd

3-Chloro-2-fluorophenol

  • MDL number MFCD01631572
  • Cas Number : 2613-22-1
  • Molecular Weight : 146.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 34-38
Purity 98
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 190
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-fluorophenol

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-fluorophenol
pictograms GHS05;GHS07;GHS09
hstatements H314;H411
Availability details for
Quantity
  • Availability
  • Add to Cart