APO455833846 Apollo Scientific Ltd

3-Chloro-4-fluorophenylacetonitrile

  • MDL number MFCD01631553
  • Cas Number : 658-98-0
  • Molecular Weight : 169.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Chloro-4-fluorobenzyl cyanide, 2-(3-Chloro-4-fluorophenyl)ethanonitrile
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorophenylacetonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorophenylacetonitrile
pictograms GHS06
hstatements H301+H311+H331;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart