APO455833857 Apollo Scientific Ltd

4-Amino-2-fluoro-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD25976371
  • Cas Number : 1448858-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Cyano-3-fluoro-2-(trifluoromethyl)aniline, 6-Amino-3-cyano-2-fluorobenzotrifluoride
Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55717

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55717
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart