APO455833858 Apollo Scientific Ltd

2-Fluoro-3-methylthiophenol

  • MDL number MFCD22481036
  • Cas Number : 1243283-09-1
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-3-methylbenzenethiol, 2-Fluoro-3-methylphenyl mercaptan
Purity 96
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-3-methylthiophenol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-3-methylthiophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart