APO455833861 Apollo Scientific Ltd

3-Chloro-2-fluoro-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD01631509
  • Cas Number : 129931-45-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-91
Synonym 3-Carboxy-5-chloro-4-fluorobenzotrifluoride, 5-Chloro-alpha,alpha,alpha,6-tetrafluoro-m-toluic acid
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2744

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2744
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart