APO455833870 Apollo Scientific Ltd

3-Bromo-5-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD09835207
  • Cas Number : 172023-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 52-54
Synonym [3-Bromo-5-(trifluoromethyl)phenyl]methanol, 3-Bromo-5-(trifluoromethyl)benzenemethanol
Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=68952

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=68952
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart