APO455833877 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-(trifluoromethyl)benzaldehyde

  • MDL number MFCD15479075
  • Cas Number : 1215310-75-0
  • Molecular Weight : 252.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-5-(methylsulphonyl)benzotrifluoride, 4-Formyl-3-(trifluoromethyl)phenyl methyl sulphone
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart