APO455833878 Apollo Scientific Ltd

4-(Methylsulphonyl)-2-(trifluoromethyl)benzyl chloride

  • MDL number MFCD11046211
  • Cas Number : 1086389-83-4
  • Molecular Weight : 272.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 91-93
Iupac Name 1-(chloromethyl)-4-(methylsulfonyl)-2-(trifluoromethyl)benzene
Inchi Code 1S/C9H8ClF3O2S/c1-16(14,15)7-3-2-6(5-10)8(4-7)9(11,12)13/h2-4H,5H2,1H3
Inchi Key PLMQGTUDXNINPI-UHFFFAOYSA-N

Description

Synonym 2-(Chloromethyl)-5-(methylsulphonyl)benzotrifluoride, 4-(Chloromethyl)-3-(trifluoromethyl)phenyl methyl sulphone
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 91-93
Product Link https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzyl-chloride

Safety Information

signalword Danger
pcodes P280;P309+P311
MSDS https://store.apolloscientific.co.uk/product/4-methylsulphonyl-2-trifluoromethylbenzyl-chloride
pictograms GHS05;GHS07
hstatements H314;H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart