APO455833881 Apollo Scientific Ltd

2'-Amino-3'-chloro-2,2,2-trifluoroacetophenone

  • MDL number MFCD22689228
  • Cas Number : 1414958-52-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(2-Amino-3-chlorophenyl)-2,2,2-trifluoroethan-1-one, 2-Chloro-6-(trifluoroacetyl)aniline
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55708

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55708
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart