APO455833882 Apollo Scientific Ltd

2'-Amino-2,2,2-trifluoro-4'-(trifluoromethyl)acetophenone

  • MDL number MFCD25976379
  • Cas Number : 1448858-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2-Amino-4-(trifluoromethyl)phenyl]-2,2,2-trifluoroethan-1-one, 2-(Trifluoroacetyl)-5-(trifluoromethyl)aniline, 3-Amino-4-(trifluoroacetyl)benzotrifluoride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=55709

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=55709
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart