APO455833884 Apollo Scientific Ltd

3-Fluoro-2-iodobenzyl alcohol

  • MDL number MFCD18396946
  • Cas Number : 1261827-82-0
  • Molecular Weight : 252.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Fluoro-2-iodophenyl)methanol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-iodobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-iodobenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart