APO455833892 Apollo Scientific Ltd

5-Chloro-2-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD01631354
  • Cas Number : 320-84-3
  • Molecular Weight : 243.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Chloro-2-(chlorocarbonyl)benzotrifluoride, 5-Chloro-alpha,alpha,alpha-trifluoro-o-toluoyl chloride
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 214-217
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-trifluoromethylbenzoyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-trifluoromethylbenzoyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart