APO455833895 Apollo Scientific Ltd

5-Chloro-2-(trifluoromethyl)benzyl alcohol

  • MDL number MFCD01631355
  • Cas Number : 261763-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-trifluoromethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-trifluoromethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart