APO455833902 Apollo Scientific Ltd

2-Amino-6-fluorobenzamide

  • MDL number MFCD00816881
  • Cas Number : 115643-59-9
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 115-118
Iupac Name 2-amino-6-fluorobenzamide
Inchi Code 1S/C7H7FN2O/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,9H2,(H2,10,11)
Inchi Key REPZELLBLWMUAB-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 2-Carbamoyl-3-fluoroaniline, 2-(Aminocarbonyl)-3-fluoroaniline
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 115-118
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-fluorobenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-fluorobenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart