APO455833903 Apollo Scientific Ltd

4-[5-(4-Fluorophenyl)-1,3,4-oxadiazol-2-yl]piperidine

  • MDL number MFCD03424488
  • Cas Number : 493024-40-1
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94-96
Synonym 2-(4-Fluorophenyl)-5-(piperidin-4-yl)-1,3,4-oxadiazole, 1-Fluoro-4-[5-(piperidin-4-yl)-1,3,4-oxadiazol-2-yl]benzene
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-5-4-fluorophenyl-134-oxadiazol-2-ylpiperidine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-5-4-fluorophenyl-134-oxadiazol-2-ylpiperidine
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart