APO455833906 Apollo Scientific Ltd

2,3-Difluoroanisole

  • MDL number MFCD01631562
  • Cas Number : 134364-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Difluoro-3-methoxybenzene, 2,3-Difluorophenyl methyl ether
Density 1.24
Flash Point 62
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/23-difluoroanisole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoroanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart