APO455833909 Apollo Scientific Ltd

2,3-Difluorobenzotrifluoride

  • MDL number MFCD01631429
  • Cas Number : 64248-59-5
  • Molecular Weight : 182.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Difluoro-3-(trifluoromethyl)benzene, alpha,alpha,alpha,2,3-Pentafluorotoluene
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluorobenzotrifluoride
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart