APO455833913 Apollo Scientific Ltd

5-Phenyl-2-(trifluoromethyl)-3-furoic acid

  • MDL number MFCD01313502
  • Cas Number : 23584-85-2
  • Molecular Weight : 256.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-166
Synonym 5-Phenyl-2-(trifluoromethyl)furan-3-carboxylic acid, 3-Carboxy-5-phenyl-2-(trifluoromethyl)furan
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-phenyl-2-trifluoromethyl-3-furoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-phenyl-2-trifluoromethyl-3-furoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart