APO455833923 Apollo Scientific Ltd

2,6-Difluoro-3-methylbenzyl bromide

  • MDL number MFCD01631336
  • Cas Number : 261763-44-4
  • Molecular Weight : 221.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Bromomethyl)-1,3-difluoro-4-methylbenzene, alpha-Bromo-2,6-difluoro-m-xylene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 76/3mm
Product Link https://store.apolloscientific.co.uk/product/26-difluoro-3-methylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS https://store.apolloscientific.co.uk/product/26-difluoro-3-methylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart