APO455833924 Apollo Scientific Ltd

2,3-Difluoro-4-methylphenol

  • MDL number MFCD01631651
  • Cas Number : 261763-45-5
  • Molecular Weight : 144.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-38
Synonym 2,3-Difluoro-p-cresol, 2,3-Difluoro-4-hydroxytoluene
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/23-difluoro-4-methylphenol

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/23-difluoro-4-methylphenol
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart