APO455833935 Apollo Scientific Ltd

5-Amino-2,4-difluorobenzotrifluoride

  • MDL number MFCD01631613
  • Cas Number : 261944-56-3
  • Molecular Weight : 197.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-24-difluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-amino-24-difluorobenzotrifluoride
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart