APO455833936 Apollo Scientific Ltd

4-Amino-2,6-difluorobenzotrifluoride

  • MDL number MFCD00190122
  • Cas Number : 123950-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66
Synonym 3,5-Difluoro-4-(trifluoromethyl)aniline, 4-Amino-alpha,alpha,alpha,2,6-pentafluorotoluene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 103-105
Product Link http://www.apolloscientific.co.uk/display_item.php?id=2275

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=2275
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart