APO455833942 Apollo Scientific Ltd

2-(4-Fluorophenyl)malonaldehyde

  • MDL number MFCD02261897
  • Cas Number : 493036-47-8
  • Molecular Weight : 166.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-167
Synonym 2-(4-Fluorophenyl)propane-1,3-dial
Purity 95
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-fluorophenylmalonaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-4-fluorophenylmalonaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart