APO455833947 Apollo Scientific Ltd

5-Bromo-1,3-dimethyl-2-fluorobenzene

  • MDL number MFCD01320701
  • Cas Number : 99725-44-7
  • Molecular Weight : 203.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-2-fluoro-m-xylene
Purity 99
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 95/22mm
Product Link https://store.apolloscientific.co.uk/product/5-bromo-13-dimethyl-2-fluorobenzene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-13-dimethyl-2-fluorobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart