APO455833949 Apollo Scientific Ltd

4-Hydroxy-2,3,5,6-tetrafluorobenzoic acid

  • MDL number MFCD00129952
  • Cas Number : 652-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148-150
Synonym 4-Carboxy-2,3,5,6-tetrafluorophenol
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-2356-tetrafluorobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-2356-tetrafluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart