APO455833956 Apollo Scientific Ltd

2,4,5-Trifluorobenzenesulphonyl chloride

  • MDL number MFCD00077600
  • Cas Number : 220227-21-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Density 1.657
Flash Point >100
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/245-trifluorobenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/245-trifluorobenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart