APO455833959 Apollo Scientific Ltd

5-Fluoro-2-methylbenzyl alcohol

  • MDL number MFCD01631527
  • Cas Number : 22062-54-0
  • Molecular Weight : 140.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36-40
Synonym (5-Fluoro-2-methylphenyl)methanol, (5-Fluoro-o-tolyl)methanol
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoro-2-methylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/5-fluoro-2-methylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart