APO455833964 Apollo Scientific Ltd

4-Fluoro-3-methylbenzyl bromide

  • MDL number MFCD01631425
  • Cas Number : 261951-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-1-fluoro-2-methylbenzene, alpha-Bromo-4-fluoro-m-xylene
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 80/3mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=5715

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=5715
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart