APO455833971 Apollo Scientific Ltd

4-Amino-3-fluorobenzotrifluoride

  • MDL number MFCD01631629
  • Cas Number : 69409-98-9
  • Molecular Weight : 179.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-4-(trifluoromethyl)aniline, alpha,alpha,alpha,2-Tetrafluoro-p-toluidine
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 55/0.3mm
Product Link https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-3-fluorobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart