APO455833972 Apollo Scientific Ltd

4-Fluoro-3-(trifluoromethyl)anisole

  • MDL number MFCD01631405
  • Cas Number : 127271-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Fluoro-5-methoxybenzotrifluoride
Purity 97
Flash Point 67
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylanisole

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-fluoro-3-trifluoromethylanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart