APO455833976 Apollo Scientific Ltd

4-Hydroxy-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD01091007
  • Cas Number : 320-32-1
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152.5-161
Synonym 4-Carboxy-3-(trifluoromethyl)phenol, 2-Carboxy-5-hydroxybenzotrifluoride
Purity 97
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-2-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart