APO455833987 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)aniline

  • MDL number MFCD01631541
  • Cas Number : 86256-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Amino-5-(trifluoromethoxy)toluene, 4-(Trifluoromethoxy)-o-toluidine
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-methyl-4-trifluoromethoxyaniline

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methyl-4-trifluoromethoxyaniline
pictograms GHS06;GHS07
hstatements H301;H311;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart