APO455833988 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)benzoic acid

  • MDL number MFCD01631542
  • Cas Number : 261951-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Synonym 2-Carboxy-5-(trifluoromethoxy)toluene, 4-(Trifluoromethoxy)-o-toluic acid
Purity 97
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4014

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4014
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart