APO455833989 Apollo Scientific Ltd

2-(4-Methoxyphenoxy)-5-(trifluoromethyl)aniline

  • MDL number MFCD00792432
  • Cas Number : 62966-74-9
  • Molecular Weight : 283.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73-75
Synonym 3-Amino-4-(4-methoxyphenoxy)benzotrifluoride, 2-Amino-4'-methoxy-4-(trifluoromethyl)diphenyl ether
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-4-methoxyphenoxy-5-trifluoromethylaniline

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/2-4-methoxyphenoxy-5-trifluoromethylaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart