APO455833990 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)benzonitrile

  • MDL number MFCD01631543
  • Cas Number : 261951-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Cyano-5-(trifluoromethoxy)toluene, 4-(Trifluoromethoxy)-o-tolunitrile
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 64
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4015

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4015
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart