APO455833991 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)benzyl alcohol

  • MDL number MFCD01631546
  • Cas Number : 261951-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym [2-Methyl-4-(trifluoromethoxy)phenyl]methanol
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4016

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4016
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart