APO455833993 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)benzyl bromide

  • MDL number MFCD01631547
  • Cas Number : 261951-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-3-methylphenyl trifluoromethyl ether, 1-(Bromomethyl)-2-methyl-4-(trifluoromethoxy)benzene
Purity 97
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4017

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4017
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart