APO455834001 Apollo Scientific Ltd

4-Methyl-2-(trifluoromethyl)benzoic acid

  • MDL number MFCD01631591
  • Cas Number : 120985-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-141
Synonym 2-Carboxy-5-methylbenzotrifluoride
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methyl-2-trifluoromethylbenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-2-trifluoromethylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart