APO455834007 Apollo Scientific Ltd

4-Methyl-2-(trifluoromethyl)benzonitrile

  • MDL number MFCD01631596
  • Cas Number : 261952-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 22-28
Synonym 2-Cyano-5-methylbenzotrifluoride, 4-Cyano-3-(trifluoromethyl)toluene, 2-(Trifluoromethyl)-p-tolunitrile
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4036

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4036
pictograms GHS06;GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart