APO455834008 Apollo Scientific Ltd

4-Methyl-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD01631597
  • Cas Number : 261952-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 5-Cyano-2-methylbenzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-methyl-3-trifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-3-trifluoromethylbenzonitrile
pictograms GHS06
hstatements H302+H332;H311;H315
Availability details for
Quantity
  • Availability
  • Add to Cart