APO455834010 Apollo Scientific Ltd

2-Methyl-5-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD01631599
  • Cas Number : 261952-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Chlorocarbonyl)-4-methylbenzotrifluoride, 3-(Chloroformyl)-4-methylbenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4042

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4042
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart