APO455834011 Apollo Scientific Ltd

4-Methyl-3-(trifluoromethyl)benzoyl chloride

  • MDL number MFCD01631602
  • Cas Number : 261952-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Chlorocarbonyl)-2-methylbenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4045

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4045
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart