APO455834018 Apollo Scientific Ltd

4-Methyl-3-(trifluoromethyl)benzyl bromide

  • MDL number MFCD01631612
  • Cas Number : 261952-19-6
  • Molecular Weight : 253.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-1-methyl-2-(trifluoromethyl)benzene, 5-(Bromomethyl)-2-methylbenzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-methyl-3-trifluoromethylbenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-methyl-3-trifluoromethylbenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart