APO455834019 Apollo Scientific Ltd

3-Bromo-2-methylbenzotrifluoride

  • MDL number MFCD04038231
  • Cas Number : 69902-83-6
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-2-methyl-3-(trifluoromethyl)benzene, 3-Bromo-alpha,alpha,alpha-trifluoro-o-xylene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 35/1mm
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-methylbenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-methylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart