APO455834020 Apollo Scientific Ltd

5-Bromo-2-methylbenzotrifluoride

  • MDL number MFCD01631587
  • Cas Number : 86845-27-4
  • Molecular Weight : 239.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-(trifluoromethyl)toluene
Purity 98
Flash Point 78
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-methylbenzotrifluoride

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-methylbenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart