APO455834021 Apollo Scientific Ltd

3,5-Difluoro-4-methoxybenzoic acid

  • MDL number MFCD01090994
  • Cas Number : 319-60-8
  • Molecular Weight : 188.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-166
Synonym 4-Carboxy-2,6-difluoroanisole, 3,5-Difluoro-p-anisic acid
Purity 99
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-difluoro-4-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-difluoro-4-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart