APO455834023 Apollo Scientific Ltd

2,3,4,5-Tetrafluorobenzyl bromide

  • MDL number MFCD01631453
  • Cas Number : 53001-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Bromomethyl)-2,3,4,5-tetrafluorobenzene, alpha-Bromo-2,3,4,5-tetrafluorotoluene
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 107-110/65mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=4789

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=4789
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart